2-Quinoxalinecarboxylic acid
Dodavatel: EHRENSTORFER GMBH
EHERC16713000EA
5740
CZK
EHERC16713000
2-Quinoxalinecarboxylic acid
2-Quinoxalinecarboxylic acid
Vzorec:
C₉H₆N₂O₂ MW: 174,16 g/mol |
MDL Number:
MFCD00012334 CAS číslo: 879-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...