Tisk…

Quinoxaline-2,3-dione, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL144789-25GEA 1550 CZK
SIAL144789-25G
Quinoxaline-2,3-dione, Sigma-Aldrich®
Quinoxaline-2,3-dione
Vzorec: C₈H₆N₂O₂
MW: 162,15 g/mol
MDL Number: MFCD00006723
CAS číslo: 15804-19-0
EINECS: 239-901-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR