Tisk…

5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde 95%

Varování

H50475.06EA 13640 CZK
H50475.06
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde 95%
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde
Vzorec: C₉H₁₁ClN₂O
MW: 158,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052538
CAS číslo: 27006-76-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR