Tisk…

4,5-Dichloro-3(2H)-pyridazinone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL303097-25GEA 0 CZK
SIAL303097-25G
4,5-Dichloro-3(2H)-pyridazinone, Sigma-Aldrich®
4,5-Dichloro-3(2H)-pyridazinone
Vzorec: C₄H₂Cl₂N₂O
MW: 164,98 g/mol
MDL Number: MFCD00051504
CAS číslo: 932-22-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR