Tisk…

1H-Indazole-6-boronic acid pinacol ester, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL692735-1GEA 0 CZK
SIAL692735-1G SIAL692735-5G
1H-Indazole-6-boronic acid pinacol ester, Sigma-Aldrich®
1H-Indazole-6-boronic acid pinacol ester
Vzorec: C₁₃H₁₇BN₂O₂
MDL Number: MFCD07368067
CAS číslo: 937049-58-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR