Tisk…

Ethyl-4-oxazolecarboxylate, Sigma-Aldrich®

Dodavatel: Merck

SIAL696625-1GEA 2340 CZK
SIAL696625-1G
Ethyl-4-oxazolecarboxylate, Sigma-Aldrich®
Ethyl-4-oxazolecarboxylate
Vzorec: C₆H₇NO₃
MW: 141,1300 g/mol
MDL Number: MFCD04114940
CAS číslo: 23012-14-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR