Tisk…

Ethyl-5-oxazolecarboxylate, Sigma-Aldrich®

Dodavatel: Merck

SIAL720763-1GEA 2320 CZK
SIAL720763-1G
Ethyl-5-oxazolecarboxylate, Sigma-Aldrich®
Ethyl-5-oxazolecarboxylate
Vzorec: C₆H₇NO₃
MW: 141,13 g/mol
MDL Number: MFCD04114930
CAS číslo: 118994-89-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR