Tisk…

2-Methyl-4-thiazolecarboxylic acid, Sigma-Aldrich®

Dodavatel: Merck

SIAL718440-1GEA 0 CZK
SIAL718440-1G
2-Methyl-4-thiazolecarboxylic acid, Sigma-Aldrich®
2-Methyl-4-thiazolecarboxylic acid
Vzorec: C₅H₅NO₂S
MW: 143,1637 g/mol
MDL Number: MFCD03407332
CAS číslo: 35272-15-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR