Tisk…

α-Bromo-2-chlorophenylacetic acid, Sigma-Aldrich®

Dodavatel: Merck

SIAL722235-25GEA 0 CZK
SIAL722235-25G
α-Bromo-2-chlorophenylacetic acid, Sigma-Aldrich®
α-Bromo-2-chlorophenylacetic acid
Vzorec: C₈H₆BrClO₂
MW: 249,49
MDL Number: MFCD03428513
CAS číslo: 29270-30-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR