4-Chloro-3-(methylcarbamoyl)phenylboronic acid ≥98%
|
Varování
|
H52659.03EA
9170
CZK
H52659.03
4-Chloro-3-(methylcarbamoyl)phenylboronic acid ≥98%
4-Chloro-3-(methylcarbamoyl)phenylboronic acid
Vzorec:
C₈H₉BClNO₃ MW: 213,43 g/mol Bod tání: 294…298 °C Storage Temperature: Ambient |
MDL Number:
MFCD07363795 CAS číslo: 871332-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...