3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
H52910.MDEA
3570
CZK
H52910.MD
3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid ≥97%
3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid
Vzorec:
C₁₀H₁₁BClNO₃ MW: 239,46 g/mol Bod tání: 216…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD07363770 CAS číslo: 850589-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...