4-Chloro-2-(trifluoromethoxy)benzaldehyde ≥97%
|
Varování
|
H33581.MDEA
1260
CZK
H33581.MD
H33581.03
4-Chloro-2-(trifluoromethoxy)benzaldehyde ≥97%
4-Chloro-2-(trifluoromethoxy)benzaldehyde
Vzorec:
C₈H₄ClF₃O₂ MW: 224,56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18393772 CAS číslo: 1261442-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...