Tisk…

Sulphaquinoxaline, Supelco®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

45662-250MGEA 3320 CZK
45662-250MG
Sulphaquinoxaline, Supelco®
Sulphaquinoxaline
Vzorec: C₁₄H₁₂N₄O₂S
MW: 300,3357 g/mol
MDL Number: MFCD00055406
CAS číslo: 59-40-5
EINECS: 200-423-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR