Tisk…

4-Methoxybenzenesulphonamide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL544655-5GEA 2220 CZK
SIAL544655-5G
4-Methoxybenzenesulphonamide, Sigma-Aldrich®
4-Methoxybenzenesulphonamide
Vzorec: C₇H₉NO₃S
MW: 187,22 g/mol
MDL Number: MFCD00025392
CAS číslo: 1129-26-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR