Tisk…

4,4'-Thiobisbenzenethiol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL396249-25GEA 0 CZK
SIAL396249-25G SIAL396249-5G
4,4'-Thiobisbenzenethiol, Sigma-Aldrich®
4,4'-Thiobisbenzenethiol
Vzorec: C₁₂H₁₀S₃
MDL Number: MFCD00142119
CAS číslo: 19362-77-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR