2,4,6-Trimethyl-N-(3-(trifluoromethyl)phenyl)benzenesulphonamide
|
Varování
|
J65999.MAEA
3280
CZK
J65999.MA
2,4,6-Trimethyl-N-(3-(trifluoromethyl)phenyl)benzenesulphonamide
2,4,6-Trimethyl-N-(3-(trifluoromethyl)phenyl)benzenesulphonamide
Activates phospholipase C
Vzorec:
C₁₆H₁₆F₃NO₂S MW: 343,36 g/mol Bod tání: 114…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00095824 CAS číslo: 200933-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...