Tisk…

4-(Benzyloxy)phenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL666920-10GEA 0 CZK
SIAL666920-10G SIAL666920-2G
4-(Benzyloxy)phenylboronic acid, Sigma-Aldrich®
4-(Benzyloxy)phenylboronic acid
Vzorec: C₁₃H₁₃BO₃
MW: 228,06
MDL Number: MFCD01075705
CAS číslo: 146631-00-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR