Tisk…

5-Boronopicolinic acid ≥97%

H53138.MDEA 4120 CZK
H53138.MD
5-Boronopicolinic acid ≥97%
5-Boronopicolinic acid
Vzorec: C₆H₆BNO₄
MW: 166,9271 g/mol
Bod tání: >300 °C
MDL Number: MFCD09027238
CAS číslo: 913836-11-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR