3-(Chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole ≥99%
H50243.MDEA
2070
CZK
H50243.MD
3-(Chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole ≥99%
3-(Chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole
Vzorec:
C₁₀H₉ClN₂O₂ MW: 224,65 g/mol Bod tání: 58…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00084966 CAS číslo: 175205-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...