2-Methyl-5-hexen-3-ol ≥99%
|
Varování
|
B20220.03EA
1100
CZK
B20220.03
B20220.14
B20220.06
2-Methyl-5-hexen-3-ol ≥99%
2-Methyl-5-hexen-3-ol
Vzorec:
C₇H₁₄O MW: 114,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00048326 CAS číslo: 32815-70-6 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...