Tisk…

(2-Methyl-3-nitrophenyl)acetic acid, Sigma-Aldrich®

Dodavatel: Merck

SIAL720216-5GEA 2420 CZK
SIAL720216-5G
(2-Methyl-3-nitrophenyl)acetic acid, Sigma-Aldrich®
(2-Methyl-3-nitrophenyl)acetic acid
Vzorec: C₉H₉NO₄
MW: 195,17 g/mol
MDL Number: MFCD07782132
CAS číslo: 23876-15-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR