Tisk…

2,2':6',2''-Terpyridine, Supelco®


M-Clarity™ quality level = MQ100

86490-250MGEA 0 CZK
86490-250MG
2,2':6',2''-Terpyridine, Supelco®
2,2':6',2''-Terpyridine
Vzorec: C₁₅H₁₁N₃
MW: 233,27 g/mol
MDL Number: MFCD00006213
CAS číslo: 1148-79-4
EINECS: 214-559-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR