Tisk…

3-Bromo-1H-indazole, Sigma-Aldrich®

Dodavatel: Merck

SIAL717525-1GEA 0 CZK
SIAL717525-1G
3-Bromo-1H-indazole, Sigma-Aldrich®
3-Bromo-1H-indazole
Vzorec: C₇H₅BrN₂
MW: 197,04 g/mol
MDL Number: MFCD00159926
CAS číslo: 40598-94-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR