3-Phenyl-5-isoxazolecarboxylic acid ≥97%
|
Varování
|
H31606.03EA
2070
CZK
H31606.03
3-Phenyl-5-isoxazolecarboxylic acid ≥97%
3-Phenyl-5-isoxazolecarboxylic acid
Vzorec:
C₁₀H₇NO₃ MW: 189,17 g/mol Bod tání: 180…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD01936010 CAS číslo: 14442-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...