Tisk…

Methyl trifluoropyruvate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL510246-5GEA 2920 CZK
SIAL510246-5G
Methyl trifluoropyruvate, Sigma-Aldrich®
Methyl trifluoropyruvate
Vzorec: C₄H₃F₃O₃
MW: 156,06 g/mol
MDL Number: MFCD00114936
CAS číslo: 13089-11-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR