8-Hexadecanol ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
B20734.22EA
0
CZK
B20734.22
8-Hexadecanol ≥97%
8-Hexadecanol
Vzorec:
C₁₆H₃₄O MW: 242,45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00046733 CAS číslo: 19781-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...