Tisk…

4-Bromotetrafluoroaniline, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL303631-5GEA 1950 CZK
SIAL303631-5G
4-Bromotetrafluoroaniline, Sigma-Aldrich®
4-Bromotetrafluoroaniline
Vzorec: C₆H₂BrF₄N
MW: 243,9844 g/mol
MDL Number: MFCD00012144
CAS číslo: 1998-66-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR