Tisk…

3-Phenoxypropiononitrile, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL493473-10GEA 2390 CZK
SIAL493473-10G
3-Phenoxypropiononitrile, Sigma-Aldrich®
3-Phenoxypropiononitrile
Vzorec: C₉H₉NO
MDL Number: MFCD00013822
CAS číslo: 3055-86-5
EINECS: 221-278-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR