Tisk…

Pentachlorothiophenol

Dodavatel: Merck

SIALMET690BEA 0 CZK
SIALMET690B
Pentachlorothiophenol
Pentachlorothiophenol
Vzorec: C₆HCl₅S
MDL Number: MFCD00059146
CAS číslo: 133-49-3
EINECS: 205-107-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR