Tisk…

2-(4-tert-Butylphenyl)ethylboronic acid pinacol ester 95%

H27051.03EA 1350 CZK
H27051.03
2-(4-tert-Butylphenyl)ethylboronic acid pinacol ester 95%
2-(4-tert-Butylphenyl)ethylboronic acid pinacol ester
Vzorec: C₁₈H₂₉BO₂
MW: 288,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953477
CAS číslo: 1073355-22-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR