Tisk…

4-Butylbenzonitrile, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL527777-5GEA 0 CZK
SIAL527777-5G
4-Butylbenzonitrile, Sigma-Aldrich®
4-Butylbenzonitrile
Vzorec: C₁₁H₁₃N
MW: 159,23
MDL Number: MFCD00052345
CAS číslo: 20651-73-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR