Tisk…

1,3,6-Trichloroisoquinoline, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL691852-1GEA 2220 CZK
SIAL691852-1G
1,3,6-Trichloroisoquinoline, Sigma-Aldrich®
1,3,6-Trichloroisoquinoline
Vzorec: C₉H₄Cl₃N
MW: 232,4900 g/mol
MDL Number: MFCD09907620
CAS číslo: 1053658-49-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR