Tisk…

4-Methoxy-2-nitrobenzonitrile, Sigma-Aldrich®

Dodavatel: Merck

SIAL579130-5GEA 0 CZK
SIAL579130-5G
4-Methoxy-2-nitrobenzonitrile, Sigma-Aldrich®
4-Methoxy-2-nitrobenzonitrile
Vzorec: C₈H₆N₂O₃
MW: 178,1500 g/mol
MDL Number: MFCD00667741
CAS číslo: 38469-83-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR