Tisk…

Methyl-4-amino-3-bromobenzoate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL655910-5GEA 9470 CZK
SIAL655910-5G
Methyl-4-amino-3-bromobenzoate, Sigma-Aldrich®
Methyl-4-amino-3-bromobenzoate
Vzorec: C₈H₈BrNO₂
MW: 230,06 g/mol
MDL Number: MFCD01861385
CAS číslo: 106896-49-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR