Fleece jacket, Match Basic
Supplier: FRISTADS KANSAS
Certificates
About this item
Anti-Pilling-fleece jacket made of 100% polyester.
- Zip to top of collar
- Two chest pockets with zip, two front pockets with zip
- Elasticated cuffs, adjustable drawstring at hem
Weight: 300 g/m²
Specifications
- Catalog No:
- KWIN1008169402XL
- KWIN100816930XS
- KWIN100816930M
- KWIN100816930S
- KWIN100816940L
- KWIN100816940M
- KWIN1008169403XL
- KWIN1008169404XL
- KWIN100816940XS
- KWIN100816940S
- KWIN1008169302XL
- KWIN1008169303XL
- Model:
- Basic
- Basic
- Basic
- Basic
- Basic
- Basic
- Basic
- Basic
- Basic
- Basic
- Basic
- Basic
- Garment application:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- Weight:
- 300 g/m²
- 300 g/m²
- 300 g/m²
- 300 g/m²
- 300 g/m²
- 300 g/m²
- 300 g/m²
- 300 g/m²
- 300 g/m²
- 300 g/m²
- 300 g/m²
- 300 g/m²
Specifications
Product Information
- Catalog #Supplier #ColourClothing SizePkAvailabilityPrice
- Supplier Article Number: 100816930XSLocal Article Number: KWIN100816930XSSpecifications:
More
Specifications
ModelBasicGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyesterWeight300 g/m² - Supplier Article Number: 100816930SLocal Article Number: KWIN100816930SSpecifications:
More
Specifications
ModelBasicGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyesterWeight300 g/m² - Supplier Article Number: 100816930MLocal Article Number: KWIN100816930MSpecifications:
More
Specifications
ModelBasicGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyesterWeight300 g/m² - Supplier Article Number: 1008169302XLLocal Article Number: KWIN1008169302XLSpecifications:
More
Specifications
ModelBasicGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyesterWeight300 g/m² - Supplier Article Number: 1008169303XLLocal Article Number: KWIN1008169303XLSpecifications:
More
Specifications
ModelBasicGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyesterWeight300 g/m² - Supplier Article Number: 100816940XSLocal Article Number: KWIN100816940XSSpecifications:
More
Specifications
ModelBasicGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyesterWeight300 g/m² - Supplier Article Number: 100816940SLocal Article Number: KWIN100816940SSpecifications:
More
Specifications
ModelBasicGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyesterWeight300 g/m² - Supplier Article Number: 100816940MLocal Article Number: KWIN100816940MSpecifications:
More
Specifications
ModelBasicGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyesterWeight300 g/m² - Supplier Article Number: 100816940LLocal Article Number: KWIN100816940LSpecifications:
More
Specifications
ModelBasicGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyesterWeight300 g/m² - Supplier Article Number: 1008169402XLLocal Article Number: KWIN1008169402XLSpecifications:
More
Specifications
ModelBasicGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyesterWeight300 g/m² - Supplier Article Number: 1008169403XLLocal Article Number: KWIN1008169403XLSpecifications:
More
Specifications
ModelBasicGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyesterWeight300 g/m² - Supplier Article Number: 1008169404XLLocal Article Number: KWIN1008169404XLSpecifications:
More
Specifications
ModelBasicGarment applicationGeneral PurposeEnvironmentally PreferableRisk areaColdGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabric100% polyesterWeight300 g/m²