Tisk…
Flame-retardant balaclava, two layers, 553 UL Spirit
Dodavatel: Fristads Kansas Group
KWIN100616940EA 1100 CZK
KWIN100616940
Flame-retardant balaclava, two layers, 553 UL Spirit
Flame-retardant balaclava made of 52% Lenzing FR® and 48% wool.
  • Covers the whole head, exposing only part of the face
  • Flame-retardant and arc tested
Weight: 230 g/m²
Certifikace: EN ISO 11612 (A1 B1 C1), EN 61482-1-2 (class 1)
OEKO-TEX® certified