Tisk…

Propargyl-p-toluenesulfonate, Sigma-Aldrich®

Dodavatel: Merck

SIAL09954-25MLEA 0 CZK
SIAL09954-25ML
Propargyl-p-toluenesulfonate, Sigma-Aldrich®
Propargyl-p-toluenesulfonate
Vzorec: C₁₀H₁₀O₃S
MW: 210,25 g/mol
MDL Number: MFCD00078365
CAS číslo: 6165-76-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR