Tisk…

1-(2-Cyanophenyl)piperazine, Sigma-Aldrich®

Dodavatel: Merck

SIAL567183-5GEA 0 CZK
SIAL567183-5G
1-(2-Cyanophenyl)piperazine, Sigma-Aldrich®
1-(2-Cyanophenyl)piperazine
Vzorec: C₁₁H₁₃N₃
MW: 187,25
MDL Number: MFCD00040793
CAS číslo: 111373-03-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR