Tisk…

2-Furoylacetonitrile, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL586528-5GEA 4050 CZK
SIAL586528-5G
2-Furoylacetonitrile, Sigma-Aldrich®
2-Furoylacetonitrile
Vzorec: C₇H₅NO₂
MW: 135,12 g/mol
MDL Number: MFCD00052210
CAS číslo: 31909-58-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR