Tisk…

3-Cyanophthalide, Sigma-Aldrich®

Dodavatel: Merck

SIAL712930-1GEA 4630 CZK
SIAL712930-1G
3-Cyanophthalide, Sigma-Aldrich®
3-Cyanophthalide
Vzorec: C₉H₅NO₂
MDL Number: MFCD15143655
CAS číslo: 27613-27-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR