Tisk…

3-Amino-5-nitrophenylboronic acid 97%

Varování

H52651.MDEA 4330 CZK
H52651.MD H52651.03
3-Amino-5-nitrophenylboronic acid 97%
3-Amino-5-nitrophenylboronic acid
Vzorec: C₆H₇BN₂O₄
MW: 181,94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08436012
CAS číslo: 89466-05-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR