Tisk…

3-Amino-5-nitrophenylboronic acid hydrochloride 96%

Dodavatel: Thermo Scientific
Varování

H52674.MDEA 10880 CZK
H52674.MD
3-Amino-5-nitrophenylboronic acid hydrochloride 96%
3-Amino-5-nitrophenylboronic acid hydrochloride
Vzorec: C₆H₈BClN₂O₄
MW: 218,40 g/mol
Bod tání: 136…141 °C
Storage Temperature: Ambient
MDL Number: MFCD03411933
CAS číslo: 389621-79-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR