Tisk…

2-Formyl-3-picoline, Sigma-Aldrich®

Dodavatel: Merck

SIAL699071-1GEA 4830 CZK
SIAL699071-1G
2-Formyl-3-picoline, Sigma-Aldrich®
2-Formyl-3-picoline
Vzorec: C₇H₇NO
MW: 121,14 g/mol
MDL Number: MFCD02181144
CAS číslo: 55589-47-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR