Tisk…

Trimellitic acid tris(2-ethylhexyl) ester, Supelco®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

92124-5MLEA 0 CZK
92124-5ML
Trimellitic acid tris(2-ethylhexyl) ester, Supelco®
Trimellitic acid tris(2-ethylhexyl) ester
Vzorec: C₃₃H₅₄O₆
MW: 546,789 g/mol
MDL Number: MFCD00053799
CAS číslo: 3319-31-1
EINECS: 222-020-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR