Tisk…

Gadolinium(III) sulphate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL575143-25GEA 5580 CZK
SIAL575143-25G
Gadolinium(III) sulphate, Sigma-Aldrich®
Gadolinium(III) sulphate
Vzorec: Gd₂O₁₂S₃
MDL Number: MFCD00011025
CAS číslo: 13628-54-1
EINECS: 237-115-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR