Tisk…

trans-2-Nitro-1-phenylpropene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL424749-5GEA 0 CZK
SIAL424749-5G
trans-2-Nitro-1-phenylpropene, Sigma-Aldrich®
trans-2-Nitro-1-phenylpropene
Vzorec: C₉H₉NO₂
CAS číslo: 18315-84-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR