Tisk…
VWR®, Standard mixture of 4 n-alkanes in ampoules for determination of oil index according to ISO9377-2
Dodavatel: Avantor
87500.010EA 10550 CZK
VWRC87500.010
VWR®, Standard mixture of 4 n-alkanes in ampoules for determination of oil index according to ISO9377-2
C10+C20+C30+C40
Concentration : 100 mg/L each
Solvent : Hexane - petroleum ether (1:1)