Tisk…
VWR®, Extraction solvent stock solution in ampoules
Dodavatel: Avantor
87501.030EA 10210 CZK
VWRC87501.030 VWRC87501.011 VWRC87501.010 VWRC87501.005
VWR®, Extraction solvent stock solution in ampoules
Mixture of C10 (20 μL/L) and C40 (20 mg/L)
Solvent: Hexane - petroleum ether (1:1)
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022