Chromatography tubing, FEP
Supplier: VICI JOUR RESEARCH
Certificates
About this item
FEP, natural or coloured.
- Solid colours identify the tubing Int.Ø.
- Recommend for ion chromatography applications
FEP (FluorinatedEthylenePropylene) tubing is chemically inert to most HPLC solvents, and is recommended for low pressure applications.
Product Information
- Catalog #Supplier #Ø int.Ext.ØColourPressureWall thicknessPkAvailabilityPrice
- JOURJR-T-6605C-M3JR-T-6605C-M30,13 mm0,79 mmRedmax. 78 bar0,33 mm3 mLog in for availability1 * 3 mSupplier Article Number: JR-T-6605C-M3Local Article Number: JOURJR-T-6605C-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6605C-M10JR-T-6605C-M100,13 mm0,79 mmRedmax. 78 bar0,33 mm10 mLog in for availability1 * 10 mSupplier Article Number: JR-T-6605C-M10Local Article Number: JOURJR-T-6605C-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6605-M1JR-T-6605-M10,13 mm0,79 mmNaturalmax. 78 bar0,33 mm1 mLog in for availability1 * 1 mSupplier Article Number: JR-T-6605-M1Local Article Number: JOURJR-T-6605-M1Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6605-M3JR-T-6605-M30,13 mm0,79 mmNaturalmax. 78 bar0,33 mm3 mLog in for availability1 * 3 mSupplier Article Number: JR-T-6605-M3Local Article Number: JOURJR-T-6605-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6605-M10JR-T-6605-M100,13 mm0,79 mmNaturalmax. 78 bar0,33 mm10 mLog in for availability1 * 10 mSupplier Article Number: JR-T-6605-M10Local Article Number: JOURJR-T-6605-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6606C-M3JR-T-6606C-M30,15 mm0,79 mmLilacmax. 78 bar0,32 mm3 mLog in for availability1 * 3 mSupplier Article Number: JR-T-6606C-M3Local Article Number: JOURJR-T-6606C-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6606C-M10JR-T-6606C-M100,15 mm0,79 mmLilacmax. 78 bar0,32 mm10 mLog in for availability1 * 10 mSupplier Article Number: JR-T-6606C-M10Local Article Number: JOURJR-T-6606C-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6606C-M25JR-T-6606C-M250,15 mm0,79 mmLilacmax. 78 bar0,32 mm25 mLog in for availability1 * 25 m20 140,00 KčSupplier Article Number: JR-T-6606C-M25Local Article Number: JOURJR-T-6606C-M25Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6606-M1JR-T-6606-M10,15 mm0,79 mmNaturalmax. 78 bar0,32 mm1 mLog in for availability1 * 1 m922,00 KčSupplier Article Number: JR-T-6606-M1Local Article Number: JOURJR-T-6606-M1Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6606-M3JR-T-6606-M30,15 mm0,79 mmNaturalmax. 78 bar0,32 mm3 mLog in for availability1 * 3 m2 630,00 KčSupplier Article Number: JR-T-6606-M3Local Article Number: JOURJR-T-6606-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6606-M10JR-T-6606-M100,15 mm0,79 mmNaturalmax. 78 bar0,32 mm10 mLog in for availability1 * 10 m8 390,00 KčSupplier Article Number: JR-T-6606-M10Local Article Number: JOURJR-T-6606-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6607C-M3JR-T-6607C-M30,18 mm0,79 mmYellowmax. 28 bar0,305 mm3 mLog in for availability1 * 3 mSupplier Article Number: JR-T-6607C-M3Local Article Number: JOURJR-T-6607C-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6607C-M10JR-T-6607C-M100,18 mm0,79 mmYellowmax. 28 bar0,305 mm10 mLog in for availability1 * 10 m8 390,00 KčSupplier Article Number: JR-T-6607C-M10Local Article Number: JOURJR-T-6607C-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6607C-M25JR-T-6607C-M250,18 mm0,79 mmYellowmax. 28 bar0,305 mm25 mLog in for availability1 * 25 mSupplier Article Number: JR-T-6607C-M25Local Article Number: JOURJR-T-6607C-M25Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6607-M1JR-T-6607-M10,18 mm0,79 mmNaturalmax. 28 bar0,305 mm1 mLog in for availability1 * 1 m922,00 KčSupplier Article Number: JR-T-6607-M1Local Article Number: JOURJR-T-6607-M1Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6607-M3JR-T-6607-M30,18 mm0,79 mmNaturalmax. 28 bar0,305 mm3 mLog in for availability1 * 3 m2 630,00 KčSupplier Article Number: JR-T-6607-M3Local Article Number: JOURJR-T-6607-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6607-M10JR-T-6607-M100,18 mm0,79 mmNaturalmax. 28 bar0,305 mm10 mLog in for availability1 * 10 m8 390,00 KčSupplier Article Number: JR-T-6607-M10Local Article Number: JOURJR-T-6607-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6610C-M3JR-T-6610C-M30,25 mm0,79 mmBluemax. 68 bar0,27 mm3 mLog in for availability1 * 3 mSupplier Article Number: JR-T-6610C-M3Local Article Number: JOURJR-T-6610C-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6610C-M10JR-T-6610C-M100,25 mm0,79 mmBluemax. 68 bar0,27 mm10 mLog in for availability1 * 10 mSupplier Article Number: JR-T-6610C-M10Local Article Number: JOURJR-T-6610C-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6610C-M25JR-T-6610C-M250,25 mm0,79 mmBluemax. 68 bar0,27 mm25 mLog in for availability1 * 25 mSupplier Article Number: JR-T-6610C-M25Local Article Number: JOURJR-T-6610C-M25Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6610-M1JR-T-6610-M10,25 mm0,79 mmNaturalmax. 68 bar0,27 mm1 mLog in for availability1 * 1 m922,00 KčSupplier Article Number: JR-T-6610-M1Local Article Number: JOURJR-T-6610-M1Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6610-M3JR-T-6610-M30,25 mm0,79 mmNaturalmax. 68 bar0,27 mm3 mLog in for availability1 * 3 m2 630,00 KčSupplier Article Number: JR-T-6610-M3Local Article Number: JOURJR-T-6610-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6610-M10JR-T-6610-M100,25 mm0,79 mmNaturalmax. 68 bar0,27 mm10 mLog in for availability1 * 10 m8 450,00 KčSupplier Article Number: JR-T-6610-M10Local Article Number: JOURJR-T-6610-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6615-M1JR-T-6615-M10,38 mm0,79 mmNaturalmax. 51 bar0,205 mm1 mLog in for availability1 * 1 m1 010,00 KčSupplier Article Number: JR-T-6615-M1Local Article Number: JOURJR-T-6615-M1Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6615-M3JR-T-6615-M30,38 mm0,79 mmNaturalmax. 51 bar0,205 mm3 mLog in for availability1 * 3 m2 930,00 KčSupplier Article Number: JR-T-6615-M3Local Article Number: JOURJR-T-6615-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6615-M10JR-T-6615-M100,38 mm0,79 mmNaturalmax. 51 bar0,205 mm10 mLog in for availability1 * 10 m9 250,00 KčSupplier Article Number: JR-T-6615-M10Local Article Number: JOURJR-T-6615-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6620-M1JR-T-6620-M10,50 mm0,79 mmNaturalmax. 35 bar0,245 mm1 mLog in for availability1 * 1 m1 020,00 KčSupplier Article Number: JR-T-6620-M1Local Article Number: JOURJR-T-6620-M1Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6620-M3JR-T-6620-M30,50 mm0,79 mmNaturalmax. 35 bar0,245 mm3 mLog in for availability1 * 3 m2 930,00 KčSupplier Article Number: JR-T-6620-M3Local Article Number: JOURJR-T-6620-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6620-M10JR-T-6620-M100,50 mm0,79 mmNaturalmax. 35 bar0,245 mm10 mLog in for availability1 * 10 m9 250,00 KčSupplier Article Number: JR-T-6620-M10Local Article Number: JOURJR-T-6620-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6620C-M3JR-T-6620C-M30,50 mm0,79 mmOrangemax. 35 bar0,245 mm3 mLog in for availability1 * 3 mSupplier Article Number: JR-T-6620C-M3Local Article Number: JOURJR-T-6620C-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6620C-M10JR-T-6620C-M100,50 mm0,79 mmOrangemax. 35 bar0,245 mm10 mLog in for availability1 * 10 mSupplier Article Number: JR-T-6620C-M10Local Article Number: JOURJR-T-6620C-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6620C-M25JR-T-6620C-M250,50 mm0,79 mmOrangemax. 35 bar0,245 mm25 mLog in for availability1 * 25 mSupplier Article Number: JR-T-6620C-M25Local Article Number: JOURJR-T-6620C-M25Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6105-M3JR-T-6105-M30,50 mm1,0 mmNaturalmax. 62 bar0,25 mm3 mLog in for availability1 * 3 m1 340,00 KčSupplier Article Number: JR-T-6105-M3Local Article Number: JOURJR-T-6105-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6105-M10JR-T-6105-M100,50 mm1,0 mmNaturalmax. 62 bar0,25 mm10 mLog in for availability1 * 10 m4 280,00 KčSupplier Article Number: JR-T-6105-M10Local Article Number: JOURJR-T-6105-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6105-M25JR-T-6105-M250,50 mm1,0 mmNaturalmax. 62 bar0,25 mm25 mLog in for availability1 * 25 m9 720,00 KčSupplier Article Number: JR-T-6105-M25Local Article Number: JOURJR-T-6105-M25Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6210-M3JR-T-6210-M31,0 mm2,0 mmNaturalmax. 62 bar0,5 mm3 mLog in for availability1 * 3 m1 340,00 KčSupplier Article Number: JR-T-6210-M3Local Article Number: JOURJR-T-6210-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6210-M10JR-T-6210-M101,0 mm2,0 mmNaturalmax. 62 bar0,5 mm10 mLog in for availability1 * 10 m4 250,00 KčSupplier Article Number: JR-T-6210-M10Local Article Number: JOURJR-T-6210-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6210-M25JR-T-6210-M251,0 mm2,0 mmNaturalmax. 62 bar0,5 mm25 mLog in for availability1 * 25 m9 630,00 KčSupplier Article Number: JR-T-6210-M25Local Article Number: JOURJR-T-6210-M25Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6310-M3JR-T-6310-M31,0 mm3,0 mmNaturalmax. 82 bar1,0 mm3 mLog in for availability1 * 3 m2 170,00 KčSupplier Article Number: JR-T-6310-M3Local Article Number: JOURJR-T-6310-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6310-M10JR-T-6310-M101,0 mm3,0 mmNaturalmax. 82 bar1,0 mm10 mLog in for availability1 * 10 m6 900,00 KčSupplier Article Number: JR-T-6310-M10Local Article Number: JOURJR-T-6310-M10Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6310-M25JR-T-6310-M251,0 mm3,0 mmNaturalmax. 82 bar1,0 mm25 mLog in for availability1 * 25 m15 620,00 KčSupplier Article Number: JR-T-6310-M25Local Article Number: JOURJR-T-6310-M25Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6320-M3JR-T-6320-M32,0 mm3,0 mmNaturalmax. 41 bar0,5 mm3 mLog in for availability1 * 3 m2 060,00 KčSupplier Article Number: JR-T-6320-M3Local Article Number: JOURJR-T-6320-M3Specifications:
Specifications
Environmentally Preferable - JOURJR-T-6320-M10JR-T-6320-M102,0 mm3,0 mmNaturalmax. 41 bar0,5 mm10 mLog in for availability1 * 10 m6 520,00 KčSupplier Article Number: JR-T-6320-M10Local Article Number: JOURJR-T-6320-M10Specifications: