Order Entry
Bonded screw caps for screw neck vials, N 15 (15-425)
Bonded screw caps for screw neck vials, N 15 (15-425)
Supplier:  MACHEREY-NAGEL
Certificates
undefined
Bonded screw caps for screw neck vials, N 15 (15-425)
Supplier:  MACHEREY-NAGEL
Product Information
  • Catalog #
    Supplier #
    Type
    Hardness
    Septum
    Thickness
    Pk
    Availability
    Price
  • MANA702114
    702114
    PP bonded, white, closed top
    55° shore A
    Silicone white/PTFE blue
    1,3 mm
    100
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS - 885,00 Kč
    885,00 Kč
    1 * 100 KS
       885,00 Kč
    Supplier Article Number: 702114
    Local Article Number: MANA702114
    Specifications:
    Specifications
    Environmentally Preferable
  • MANA702180
    702180
    PP bonded, black, center hole
    45° shore A
    Silicone white/PTFE beige
    1,5 mm
    100
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS - 1 840,00 Kč
    1 840,00 Kč
    1 * 100 KS
       1 840,00 Kč
    Supplier Article Number: 702180
    Local Article Number: MANA702180
    Specifications:
    Specifications
    Environmentally Preferable